Learn More
Medchemexpress LLC 2-Morpholinoethanesulphonic acid monohydrate | 145224-94-8 | 99.95% | 250 MG
SDP
Catalog No. 5000414440
Encompass_Preferred
MES monohydrate is an analytical standard intended for research and analytical applications. It is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments as a reference standard for assays.
  • Used in cell culture
  • Used in enzyme activity determination
  • Used in electrophoresis
  • Used in protein studies
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-14440 Supplier Medchemexpress LLC Supplier No. 250MGHYD0858AR
MES monohydrate is an analytical standard intended for research and analytical applications. It is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments as a reference standard for assays.
  • Used in cell culture
  • Used in enzyme activity determination
  • Used in electrophoresis
  • Used in protein studies
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS