Learn More
Medchemexpress LLC 2-Morpholinoethanesulphonic acid monohydrate (Standard) | 145224-94-8 | 99.95% | 50 MG
SDP
Catalog No. 5000410652
Encompass_Preferred
MES monohydrate (Standard) is the analytical standard of MES (monohydrate), intended for research and analytical applications. MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments.
  • Used for research and analytical applications.
  • Amphoteric ion buffer.
  • Buffer capacity ranging pH 5.5-7.0.
  • Widely used in biochemistry and molecular biology experiments.
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-10652 Supplier Medchemexpress LLC Supplier No. HYD0858AR50MG
MES monohydrate (Standard) is the analytical standard of MES (monohydrate), intended for research and analytical applications. MES (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. As a Good's buffer, it is widely used in biochemistry and molecular biology experiments.
  • Used for research and analytical applications.
  • Amphoteric ion buffer.
  • Buffer capacity ranging pH 5.5-7.0.
  • Widely used in biochemistry and molecular biology experiments.
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.