Learn More
Medchemexpress LLC 2-Morpholinoethanesulphonic acid potassium | 39946-25-3 | 99.9% | 50 G
SDP
Catalog No. 5000409960
Encompass_Preferred
MES potassium is an amphoteric ion buffer widely used in biochemistry and molecular biology experiments. It provides effective buffering capacity around pH 6.15 and is suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Amphoteric ion buffer
  • Features a buffer capacity ranging from pH 5.5-7.0
  • Functions as a zwitterionic buffer
  • Exhibits good buffering properties in biological experiments
  • Shows poor affinity with metal ions, suitable for metal ion studies
Catalog No. 50-004-09960 Supplier Medchemexpress LLC Supplier No. HYD0858E50G
Add to Cart
Add to Cart
MES potassium is an amphoteric ion buffer widely used in biochemistry and molecular biology experiments. It provides effective buffering capacity around pH 6.15 and is suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Amphoteric ion buffer
  • Features a buffer capacity ranging from pH 5.5-7.0
  • Functions as a zwitterionic buffer
  • Exhibits good buffering properties in biological experiments
  • Shows poor affinity with metal ions, suitable for metal ion studies