Learn More
AdipoGen 2-Morpholinoethyl isocyanide
SDP
Catalog No. 502560449
Encompass_Preferred
Chemical. CAS 78375-48-1. Formula C7H12N2O. MW BD9837. 2-Morpholinoethyl isocyanide is a buildig block and intermediate for organic synthesis. The principal synthetic application of 2-morpholinoethyl isocyanide is as a coupling reagent of amino acids to yield peptides. It is a potent coupling reagent for peptide synthesis giving higher yields and less racemization than DCCI. It also reacts with aldehydes or ketones to yield amides, with acids to yield imidazolinium salts, and has been used as a ligand in spectroscopic studies to examine shielding effects. 2-Morpholinoethyl isocyanide can be used for the synthesis of nitrogen acyclic carbene NAC complexes to be used as catalysts for the synthesis of phenol and hydration of alkynes. It can also be used as a ligand in bis-cyclometalated iridium complexes for the organic light emitting diodes OLEDs.
Catalog No. 50-256-0449 Supplier AdipoGen Supplier No. CDXM0701L010
Chemical. CAS 78375-48-1. Formula C7H12N2O. MW BD9837. 2-Morpholinoethyl isocyanide is a buildig block and intermediate for organic synthesis. The principal synthetic application of 2-morpholinoethyl isocyanide is as a coupling reagent of amino acids to yield peptides. It is a potent coupling reagent for peptide synthesis giving higher yields and less racemization than DCCI. It also reacts with aldehydes or ketones to yield amides, with acids to yield imidazolinium salts, and has been used as a ligand in spectroscopic studies to examine shielding effects. 2-Morpholinoethyl isocyanide can be used for the synthesis of nitrogen acyclic carbene NAC complexes to be used as catalysts for the synthesis of phenol and hydration of alkynes. It can also be used as a ligand in bis-cyclometalated iridium complexes for the organic light emitting diodes OLEDs.