Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Morpholinoethyl isocyanide

Catalog No. 6989350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-893-50ML 50 mL
69-893-10ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-893-50ML Supplier Sigma Aldrich Supplier No. 6989350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-morpholinoethyl isocyanide can be used for the synthesis of:
  • Nitrogen acyclic carbene (nac) complexes to be used as catalysts for the synthesis of phenol and hydration of alkynes
  • Variety of imidazo[1,2-a]pyridines and α-acylaminoamides by ugi condensation reaction
  • 2-aminobenzoxazoles and 3-aminobenzoxazines having important therapeutic activities.it can also be used as a ligand in bis-cyclometalated iridium complexes for the organic light emitting diodes (oleds).potent coupling reagent for peptide synthesis giving higher yields and less racemization than dcci; reagent used in a passerini-like reaction to alpha-hydroxy amides

Specifications

CAS 78375-48-1
Density 1.017 g/mL (at 20°C (literature))
Boiling Point 72°C to 73°C (0.7 mmHg)
Molecular Formula C7H12N2O
Refractive Index n20/D 1.469
Linear Formula C7H12N2O
MDL Number MFCD00040506
Quantity 50 mL
Synonym 4-(2-Isocyanoethyl)morpholine; MEI
Molecular Weight (g/mol) 140.18
Percent Purity ≥98.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.