Learn More
Sigma Aldrich 2-N-Boc-amino-thiazole-5-carboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Claisen type reaction of n,n-dimethylacetamide dimethyl acetal using optically active trans-3-penten-2-ol as substrate has been investigated.n,n-dimethylacetamide dimethyl acetal has been used as:
- One carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones
- Reagent for the synthesis of amides, diacylamines and heterocycles
Specifications
Specifications
| Molecular Formula | C9H12N2O4S |
| Linear Formula | C9H12N2O4S |
| MDL Number | MFCD06796614 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 244.27 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.