Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Butylfuran, 98%

Catalog No. AAA1337609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337609 10 g
AAA1337618 50 g
AAA1337630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1337609 Supplier Thermo Scientific Chemicals Supplier No. A1337609
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4466-24-4 | C8H12O | 124.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4466-24-4
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00047071
InChI Key NWZIYQNUCXUJJJ-UHFFFAOYSA-N
Synonym furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane
PubChem CID 20534
IUPAC Name 2-butylfuran
SMILES CCCCC1=CC=CO1

Specifications

Density 0.886
Boiling Point 153°C to 154°C
Flash Point 32°C (89°F)
Odor Pleasant
Quantity 10 g
UN Number UN1993
Beilstein 107045
Formula Weight 124.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-n-Butylfuran
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement

H226

Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 224-732-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.