Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Dodecylfuran, MP Biomedicals™

Catalog No. MP219375391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219375391 1 g
MP219375380 100 mg
MP219375390 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219375391 Supplier MP Biomedicals, Inc Supplier No. 0219375391
Add to Cart
Add to Cart

2-n-Dodecylfuran Clear, Colorless Liquid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75308-12-2
Molecular Formula C16H28O
Molecular Weight (g/mol) 236.40
InChI Key YCHOIEWPPPNUFM-UHFFFAOYSA-N

Specifications

Quantity 1 g
Physical Form Liquid
Assay 0.98
storageNote1 +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.