Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(N-Ethylanilino)ethanol 98.0+%, TCI America™
SDP

Catalog No. E006225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006225G 25 g
E0062500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E006225G Supplier TCI America Supplier No. E006225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92-50-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00020575
InChI Key HYVGFUIWHXLVNV-UHFFFAOYSA-N
Synonym 2-n-ethylanilino ethanol, 2-ethyl phenyl amino ethanol, ethanol, 2-ethylphenylamino, ethylphenylethanolamine, phenylethylethanolamine, n-ethyl-n-2-hydroxyethyl aniline, n-ethylanilinoethanol, hydroxyethylethylaniline, n-ethyl-n-hydroxyethylaniline, 2-ethylphenylamino ethanol
PubChem CID 62338
IUPAC Name 2-[ethyl(phenyl)amino]ethan-1-ol
SMILES CCN(CCO)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 268°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(N-Ethylanilino)ethanol
EINECSNumber (3)-0159
RTECSNumber KL0878000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.