Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(N-Methyl-2-pyridylamino)ethanol 98.0+%, TCI America™
SDP

Catalog No. M14685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14685G Supplier TCI America Supplier No. M14685G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122321-04-4
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD02667633
InChI Key MWGKOPUDDQZERY-UHFFFAOYSA-N
Synonym 2-methyl pyridin-2-yl amino ethanol, 2-methyl-2-pyridylamino ethanol, 2-methyl pyridin-2-yl amino ethan-1-ol, 2-n-methyl-2-pyridylaminoethanol, 2-methyl-2-pyridinylamino ethanol, 2-n-methyl-2-pyridylamino ethanol, 2-n-methyl-n-2-pyridinyl amino ethanol, 2-n-2-hydroxyethyl-n-methylamino pyridine, 2-methyl 2-pyridinyl amino-1-ethanol
PubChem CID 737420
IUPAC Name 2-[methyl(pyridin-2-yl)amino]ethanol
SMILES CN(CCO)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 152.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(N-Methyl-2-pyridylamino)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.