Learn More
Strem, An Ascensus Company CAS# 415941-58-1. 1g. [2-(N,N-Dimethylamino)phenyl]di-t-butylphosphine, min. 95%.
SDP
Supplier:  Strem, An Ascensus Company 1512601G
Encompass
CAS# 415941-58-1. 1g. [2-(N,N-Dimethylamino)phenyl]di-t-butylphosphine, min. 95%. . Molecular Weight: 265.37. Molecular Formula: C16H28NP. Color/form: white to light-brown xtl. Strem# 15-1260. http://www.strem.com/catalog/v/15-1260/
Catalog No. 50-138-5406