Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Octyl-1-dodecylamine 97.0+%, TCI America™
SDP

Catalog No. O04351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04351G Supplier TCI America Supplier No. O04351G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62281-06-5
Molecular Formula C20H43N
Molecular Weight (g/mol) 297.571
MDL Number MFCD19440743
InChI Key VDNQHHBRKZQBPY-UHFFFAOYSA-N
Synonym 9-(Aminomethyl)nonadecane, 2-Octyldodecan-1-amine
PubChem CID 18761999
IUPAC Name 2-octyldodecan-1-amine
SMILES CCCCCCCCCCC(CCCCCCCC)CN

Specifications

Color Yellow
Quantity 1 g
UN Number 2735
Formula Weight 297.57
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-n-Octyl-1-dodecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.