Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Octyl-4-isothiazolin-3-one 98.0+%, TCI America™
SDP

Catalog No. O034325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O034325G 25 g
O03435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O034325G Supplier TCI America Supplier No. O034325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26530-20-1
Molecular Formula C11H19NOS
Molecular Weight (g/mol) 213.339
MDL Number MFCD00072473
InChI Key JPMIIZHYYWMHDT-UHFFFAOYSA-N
Synonym Octhilinone
PubChem CID 33528
ChEBI CHEBI:81936
IUPAC Name 2-octyl-1,2-thiazol-3-one
SMILES CCCCCCCCN1C(=O)C=CS1

Specifications

Boiling Point 142°C
Quantity 25 g
UN Number 2923
Formula Weight 213.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-n-Octyl-4-isothiazolin-3-one
EINECSNumber (5)-5246
RTECSNumber NX8156900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.