Learn More
Medchemexpress LLC 2-naphthalen-1,3,4,5,6,7,8-d7-amine | 93951-94-1 | 98.0% | 150.23 g/mol | C10D7NH2 | 10 MG
SDP
Catalog No. 5000281070
Encompass_Preferred
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled analogue of 2-aminonaphthalene used as a stable isotope internal standard and tracer in analytical chemistry. It is supplied as a powder for laboratory use and can be dissolved for analytical workflows.
Catalog No. 50-002-81070 Supplier Medchemexpress LLC Supplier No. HYW450322S10MG
Add to Cart
Add to Cart
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled analogue of 2-aminonaphthalene used as a stable isotope internal standard and tracer in analytical chemistry. It is supplied as a powder for laboratory use and can be dissolved for analytical workflows.