Learn More
Medchemexpress LLC 2-naphthalen-1,3,4,5,6,7,8-d7-amine | 93951-94-1 | MFCD00142864 | 99.0% | 150.23 g/mol | C10H2D7N | 1 MG
SDP
Catalog No. 5000281030
Encompass_Preferred
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled isotopologue of 2-aminonaphthalene supplied as a powdered laboratory reagent for research use. It is used as an internal standard, tracer, or labeled building block in analytical and synthetic applications; follow manufacturer handling and storage guidelines.
  • Deuterium-labeled isotopologue suitable for tracer and analytical studies.
  • High purity (99.0%) for consistent analytical performance.
  • Powder form with recommended cold storage to maintain stability.
  • Molecular formula C10H2D7N and molecular weight 150.23 g/mol.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-81030 Supplier Medchemexpress LLC Supplier No. HYW450322S1MG
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled isotopologue of 2-aminonaphthalene supplied as a powdered laboratory reagent for research use. It is used as an internal standard, tracer, or labeled building block in analytical and synthetic applications; follow manufacturer handling and storage guidelines.
  • Deuterium-labeled isotopologue suitable for tracer and analytical studies.
  • High purity (99.0%) for consistent analytical performance.
  • Powder form with recommended cold storage to maintain stability.
  • Molecular formula C10H2D7N and molecular weight 150.23 g/mol.
  • Intended for research use only; not for human or clinical applications.