Learn More
Medchemexpress LLC 2-naphthalen-1,3,4,5,6,7,8-d7-amine | 93951-94-1 | MFCD00142864 | 99.0% | 150.23 g/mol | C10H2ND7 | 5 MG
SDP
Catalog No. 5000281479
Encompass_Preferred
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled form of 2-aminonaphthalene supplied for laboratory research use as an isotopically labeled reference standard. It is intended for analytical applications such as mass spectrometry, tracer experiments, and metabolic studies; handle and store according to the product SDS and supplier recommendations.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic enrichment suitable for quantitative analysis.
  • Available in small milligram pack sizes for analytical workflows.
  • Stable as a powder under low-temperature storage when handled per SDS instructions.
  • Commonly used in tracer, metabolic, and pharmacokinetic studies.
Catalog No. 50-002-81479 Supplier Medchemexpress LLC Supplier No. HYW450322S5MG
2-Naphthalen-1,3,4,5,6,7,8-d7-amine is the deuterium-labeled form of 2-aminonaphthalene supplied for laboratory research use as an isotopically labeled reference standard. It is intended for analytical applications such as mass spectrometry, tracer experiments, and metabolic studies; handle and store according to the product SDS and supplier recommendations.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic enrichment suitable for quantitative analysis.
  • Available in small milligram pack sizes for analytical workflows.
  • Stable as a powder under low-temperature storage when handled per SDS instructions.
  • Commonly used in tracer, metabolic, and pharmacokinetic studies.