Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Naphthalen-1-yl)-5-phenyloxazole | 846-63-9 | 99.7% | 271.31 g/mol | C19H13NO | 1 G
SDP

Catalog No. 5000208925
Encompass_Preferred

2-(Naphthalen-1-yl)-5-phenyloxazole (α-NPO) is an organic fluorescent dye supplied as an off-white to light yellow solid. It has molecular formula C19H13NO, a molecular weight of 271.31 g/mol, and a reported purity of 99.73%. Recommended storage is solid at -20°C protected from light; solutions have defined shelf-life at low temperatures.

  • High purity: 99.7% (reported)
  • Physical form: solid, off-white to light yellow
  • Storage: solid at -20°C, protect from light; in solvent store at -80°C up to 6 months
  • Application: organic fluorescent dye for research and analytical assays
  • Cas number: 846-63-9

Catalog No. 50-002-08925 Supplier Medchemexpress LLC Supplier No. HYW0402091G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Naphthalen-1-yl)-5-phenyloxazole (α-NPO) is an organic fluorescent dye supplied as an off-white to light yellow solid. It has molecular formula C19H13NO, a molecular weight of 271.31 g/mol, and a reported purity of 99.73%. Recommended storage is solid at -20°C protected from light; solutions have defined shelf-life at low temperatures.

  • High purity: 99.7% (reported)
  • Physical form: solid, off-white to light yellow
  • Storage: solid at -20°C, protect from light; in solvent store at -80°C up to 6 months
  • Application: organic fluorescent dye for research and analytical assays
  • Cas number: 846-63-9

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.