Learn More
Medchemexpress LLC 2-(Naphthalen-1-yl)ethanol | 773-99-9 | MFCD00004050 | 99.9% | 172.22 | C12H12O | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01687650G
Encompass_Preferred
2-(Naphthalen-1-yl)ethanol (CAS 773-99-9) is an organic research chemical commonly used as a building block and reagent in medicinal chemistry and small-molecule synthesis. It is supplied as a white to off-white solid and is provided with analytical characterization and high reported purity.
  • High purity suitable for research (99.88% by HPLC).
  • White to off-white solid form for ease of handling.
  • Molecular formula C12H12O and molecular weight 172.22.
  • Useful as a synthetic building block and intermediate in organic synthesis.
  • Characterized by standard analytical data (HPLC, NMR).
Catalog No. 50-003-01055