Learn More
Medchemexpress LLC 2-(naphthalen-2-yl)acetic acid | 581-96-4 | MFCD00004126 | 99.9% | 186.21 g/mol | C12H10O2 | 10 G
SDP
Catalog No. 5000302278
Encompass_Preferred
2-(Naphthalen-2-yl)acetic acid is an organic small molecule used as a plant growth regulator and as a biochemical probe to study amino acid conjugation mechanisms. Supplied as a solid powder for research use, it has high reported purity and defined cold-storage recommendations; sales may be restricted in certain territories.
  • Plant growth regulator that can improve crop quality and yield.
  • Useful as a biochemical probe to study amino acid conjugation mechanisms.
  • Supplied as a solid powder with recommended cold storage for stability.
  • High purity suitable for analytical and biological research applications.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-003-02278 Supplier Medchemexpress LLC Supplier No. HYW01618710G
2-(Naphthalen-2-yl)acetic acid is an organic small molecule used as a plant growth regulator and as a biochemical probe to study amino acid conjugation mechanisms. Supplied as a solid powder for research use, it has high reported purity and defined cold-storage recommendations; sales may be restricted in certain territories.
  • Plant growth regulator that can improve crop quality and yield.
  • Useful as a biochemical probe to study amino acid conjugation mechanisms.
  • Supplied as a solid powder with recommended cold storage for stability.
  • High purity suitable for analytical and biological research applications.
  • Intended for research use only; not for human therapeutic use.