Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(naphthalen-2-yl)acetic acid | 581-96-4 | MFCD00004126 | 99.9% | 186.21 g/mol | C12H10O2 | 25 G
SDP

Catalog No. 5000304199
Encompass_Preferred

2-(Naphthalen-2-yl)acetic acid (2-naphthylacetic acid) is a plant growth regulator and research reagent used to modulate plant development and to probe amino acid conjugation mechanisms in biological studies. Supplied as a solid at analytical-grade purity, it is suitable for laboratory and agricultural research applications.

  • CAS 581-96-4
  • Molecular formula C12H10O2
  • Molecular weight 186.21 g/mol
  • High purity (up to 99.9%)
  • Available pack sizes: 5 g, 10 g, 25 g, 50 g
  • Solid form suitable for biochemical and plant studies

Catalog No. 50-003-04199 Supplier Medchemexpress LLC Supplier No. 25GHYW016187
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Naphthalen-2-yl)acetic acid (2-naphthylacetic acid) is a plant growth regulator and research reagent used to modulate plant development and to probe amino acid conjugation mechanisms in biological studies. Supplied as a solid at analytical-grade purity, it is suitable for laboratory and agricultural research applications.

  • CAS 581-96-4
  • Molecular formula C12H10O2
  • Molecular weight 186.21 g/mol
  • High purity (up to 99.9%)
  • Available pack sizes: 5 g, 10 g, 25 g, 50 g
  • Solid form suitable for biochemical and plant studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.