Learn More
Medchemexpress LLC 2-(naphthalen-2-yl)acetic acid | 581-96-4 | MFCD00004126 | 99.9% | 186.21 g/mol | C12H10O2 | 5 G
SDP
Catalog No. 5000302168
Encompass_Preferred
2-(naphthalen-2-yl)acetic acid is a synthetic auxin used as a plant growth regulator and research reagent. It is supplied as a white to off-white solid with high purity, suitable for laboratory and agricultural studies.
  • High purity (99.9%) for reliable experimental outcomes
  • Solid form, white to off-white, easy to weigh and handle
  • Suitable for auxin-related plant growth and physiology research
  • Chemical formula C12H10O2; molecular weight 186.21 g/mol
  • Available in small packaging for research-scale use
Catalog No. 50-003-02168 Supplier Medchemexpress LLC Supplier No. HYW0161875G
2-(naphthalen-2-yl)acetic acid is a synthetic auxin used as a plant growth regulator and research reagent. It is supplied as a white to off-white solid with high purity, suitable for laboratory and agricultural studies.
  • High purity (99.9%) for reliable experimental outcomes
  • Solid form, white to off-white, easy to weigh and handle
  • Suitable for auxin-related plant growth and physiology research
  • Chemical formula C12H10O2; molecular weight 186.21 g/mol
  • Available in small packaging for research-scale use