Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(naphthalen-2-yl)acetonitrile | 7498-57-9 | MFCD00004122 | 167.21 g/mol | C12H9N | 25 G
SDP

Catalog No. 5000303489
Encompass_Preferred

2-(Naphthalen-2-yl)acetonitrile (CAS 7498-57-9) is an organic intermediate used in pharmaceutical and chemical research to build more complex molecules. It is a small, nitrile-bearing aromatic compound with formula C12H9N and molecular weight 167.21 g/mol, supplied for laboratory research use only.

  • Used as a building block for synthesis of active compounds.
  • Molecular formula C12H9N.
  • Molecular weight 167.21 g/mol.
  • Provided as a solid reagent suitable for organic synthesis.
  • Intended for research use only; not for human or clinical use.
  • Available in laboratory pack sizes including 25 g.

Catalog No. 50-003-03489 Supplier Medchemexpress LLC Supplier No. 25GHYW017116
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Naphthalen-2-yl)acetonitrile (CAS 7498-57-9) is an organic intermediate used in pharmaceutical and chemical research to build more complex molecules. It is a small, nitrile-bearing aromatic compound with formula C12H9N and molecular weight 167.21 g/mol, supplied for laboratory research use only.

  • Used as a building block for synthesis of active compounds.
  • Molecular formula C12H9N.
  • Molecular weight 167.21 g/mol.
  • Provided as a solid reagent suitable for organic synthesis.
  • Intended for research use only; not for human or clinical use.
  • Available in laboratory pack sizes including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.