Learn More
Medchemexpress LLC 2-Naphthalenemethanol | 1592-38-7 | MFCD00004074 | 158.20 | 1 ML
SDP
Catalog No. 5000446122
Encompass_Preferred
2-Naphthalenemethanol is the metabolite of the environmental pollutant 2-methylnaphthalene. It covalently binds to alveolar protein and induces pulmonary toxicity. This product is supplied as a 10 mM solution in DMSO and is intended for research use only.
  • Formulated as a 10 mM solution in DMSO
  • For research use only
  • Store at -80°C for up to 6 months or -20°C for 1 month (in solvent)
Catalog No. 50-004-46122 Supplier Medchemexpress LLC Supplier No. HYW00195310MM/1ML
2-Naphthalenemethanol is the metabolite of the environmental pollutant 2-methylnaphthalene. It covalently binds to alveolar protein and induces pulmonary toxicity. This product is supplied as a 10 mM solution in DMSO and is intended for research use only.
  • Formulated as a 10 mM solution in DMSO
  • For research use only
  • Store at -80°C for up to 6 months or -20°C for 1 month (in solvent)