Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Naphthalenesulfonic acid

Catalog No. 249548100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-954-8100G 100 g
24-954-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-954-8100G Supplier Sigma Aldrich Supplier No. 249548100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The fluorescence of 2-naphthalenesulfonic acid (nsa) is effectively quenched by colloidal silver via photoinduced interfacial electron transfer from the excited singlet of nsa to silver
  • Adsorption behavior of nsa on a conventional macroporous resin amberlite xad4 and hypercrosslinked resins nda101 and nda100 has been investigated

Specifications

CAS 120-18-3
Molecular Formula C10H8O3S
Linear Formula C10H7SO3H
MDL Number MFCD00004089
Quantity 100 g
Molecular Weight (g/mol) 208.23
Percent Purity 70%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.