Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthoic hydrazide, 98%, Thermo Scientific™

Catalog No. AAL0803806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0803806 5 g
AAL0803803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0803806 Supplier Thermo Scientific Chemicals Supplier No. L0803806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39627-84-4
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD00016810
InChI Key RARLPRMZJNIQGU-UHFFFAOYSA-N
Synonym 2-naphthohydrazide, 2-naphthhydrazide, 2-naphthoic hydrazide, 2-naphthalenecarboxylic acid, hydrazide, .beta.-naphthoylhydrazine, naphthalene-2-carboxylic acid hydrazide, zlchem 976, 2-naphthohydrazide #, beta-naphthoylhydrazine, acmc-1afu5
PubChem CID 123485
IUPAC Name naphthalene-2-carbohydrazide
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)NN

Specifications

Melting Point 145°C to 149°C
Boiling Point 302°C to 304°C (725 mmHg)
Quantity 5 g
Beilstein 1950982
Formula Weight 186.21
Percent Purity 98%
Chemical Name or Material 2-Naphthoic hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-547-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.