Learn More
Medchemexpress LLC 2-naphthol | 135-19-3 | MFCD00004067 | 99.9% | 144.17 g·mol⁻¹ | C10H8O | 250g
SDP
Catalog No. 5000268379
Encompass_Preferred
2-Naphthol is a metabolite of naphthalene catalyzed by cytochrome P450 (CYP) isozymes (CYP 1A1 CYP 1A2 CYP 2A1 CYP 2E1 and CYP 2F2)
Catalog No. 50-002-68379 Supplier Medchemexpress LLC Supplier No. HYY0110250G
2-Naphthol is a metabolite of naphthalene catalyzed by cytochrome P450 (CYP) isozymes (CYP 1A1 CYP 1A2 CYP 2A1 CYP 2E1 and CYP 2F2)