Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-naphthol-d7 | 78832-54-9 | MFCD01074243 | 95.0% | 151.22 g/mol | C10HD7O | 100mg
SDP

Catalog No. 5000270067
Encompass_Preferred

2-Naphthol-d7 is the deuterium labeled 2-Naphthol[1] 2-Naphthol is a metabolite of naphthalene catalyzed by cytochrome P450 (CYP) isozymes (CYP 1A1 CYP 1A2 CYP 2A1 CYP 2E1 and CYP 2F2)[2]

Catalog No. 50-002-70067 Supplier Medchemexpress LLC Supplier No. HYY0110S1100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Naphthol-d7 is the deuterium labeled 2-Naphthol[1] 2-Naphthol is a metabolite of naphthalene catalyzed by cytochrome P450 (CYP) isozymes (CYP 1A1 CYP 1A2 CYP 2A1 CYP 2E1 and CYP 2F2)[2]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.