Learn More
Bioworld 2-Naphthoxyacetic Acid, 100 g
Supplier:  Bioworld 306310041
Encompass_Preferred
2-Naphthoxyacetic acidSynonyms: β-Napthoxyacetic acid, BNOAInduces root proliferation and prevents fruit from falling prematurelyPlant tissue culture testedAppearance: grey crystalMelting Point: 151-154°CPartially soluble in water; Soluble in DMSOProtect from moisture
Catalog No. 50-255-034