Learn More
Medchemexpress LLC 2-Naphthoxyacetic acid | 120-23-0 | MFCD00004066 | 99.9% | 202.21 | C12H10O3 | 1 ML
SDP
Catalog No. 5000303791
Encompass_Preferred
2-Naphthoxyacetic acid is an endogenous metabolite supplied for laboratory research use. It has formula C12H10O3, molecular weight 202.21, and is reported with high purity; available as a solution in DMSO and as a solid for custom use.
  • High reported purity (99.89%), suitable for analytical and preparative studies.
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL) for convenient dosing.
  • Also supplied as solid for long-term storage or custom formulation.
  • Datasheet and certificate of analysis available to support quality assurance.
  • Recommended storage: powder at -20°C; stock solutions at -80°C for extended stability.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-003-03791 Supplier Medchemexpress LLC Supplier No. HYW01479610MM/1ML
2-Naphthoxyacetic acid is an endogenous metabolite supplied for laboratory research use. It has formula C12H10O3, molecular weight 202.21, and is reported with high purity; available as a solution in DMSO and as a solid for custom use.
  • High reported purity (99.89%), suitable for analytical and preparative studies.
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL) for convenient dosing.
  • Also supplied as solid for long-term storage or custom formulation.
  • Datasheet and certificate of analysis available to support quality assurance.
  • Recommended storage: powder at -20°C; stock solutions at -80°C for extended stability.
  • Intended for research use only; not for human or clinical applications.