Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl Laurate 98.0+%, TCI America™
SDP

Catalog No. L012225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L012225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L012225G Supplier TCI America Supplier No. L012225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6343-73-3
Molecular Formula C22H30O2
Molecular Weight (g/mol) 326.48
MDL Number MFCD00021608
InChI Key CIGGUBXZFFSTTA-UHFFFAOYSA-N
Synonym Lauric Acid 2-Naphthyl Ester
PubChem CID 80656
IUPAC Name naphthalen-2-yl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 61°C
Quantity 25 g
Formula Weight 326.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Laurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.