Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthylamine, MilliporeSigma™ Supelco™

Catalog No. 111019950 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9950 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9950 Supplier MilliporeSigma Supelco Supplier No. 31618100MG
Add to Cart
Add to Cart

2-Naphthylamine is a carcinogenic aromatic amine which induces tumor in urinary bladder.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 91-59-8
Density 1.061 g/mL (at 25°C)
Physical Form Neat
Linear Formula C10H7 NH2
MDL Number MFCD00004112
Melting Point 111°C to 113°C
Boiling Point 306°C
Synonym beta-Naphthylamine; 2-Aminonaphthalene
Type Analytical Standard
Quantity 100 mg
Formula Weight 143.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.