Learn More
Medchemexpress LLC 2-Naphthylboronic acid | 32316-92-0 | 171.99 | 100 MG
SDP
Catalog No. 5000445839
Encompass_Preferred
2-Naphthylboronic acid is a biochemical reagent suitable for life science related research. It can be used as a biological material or an organic compound.
  • Undergoes silylation reaction
  • Readily participates in Suzuki-Miyaura cross-coupling
Catalog No. 50-004-45839 Supplier Medchemexpress LLC Supplier No. HYW002570S100MG
2-Naphthylboronic acid is a biochemical reagent suitable for life science related research. It can be used as a biological material or an organic compound.
  • Undergoes silylation reaction
  • Readily participates in Suzuki-Miyaura cross-coupling