Learn More
Medchemexpress LLC 2-Naphthylboronic acid | 32316-92-0 | 171.99 | 25 G
SDP
Catalog No. 5000448113
Encompass_Preferred
2-Naphthylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is available in solid form with a white to off-white appearance. This compound has several key properties that make it valuable in research.
  • Can undergo silylation reaction.
  • Readily undergoes Suzuki-Miyaura cross-coupling.
  • Suitable for life science related research.
Catalog No. 50-004-48113 Supplier Medchemexpress LLC Supplier No. 25GHYW002570
2-Naphthylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is available in solid form with a white to off-white appearance. This compound has several key properties that make it valuable in research.
  • Can undergo silylation reaction.
  • Readily undergoes Suzuki-Miyaura cross-coupling.
  • Suitable for life science related research.