Learn More
Medchemexpress LLC 2-Naphthylboronic acid | 32316-92-0 | 171.99 | 5 G

Supplier: Medchemexpress LLC HYW002570S5G
2-Naphthylboronic acid is a biochemical reagent used in life science research. It can undergo silylation reaction and readily undergoes Suzuki-Miyaura cross-coupling, making it valuable as a biological material or organic compound.
- Biochemical reagent for life science research
- Undergoes silylation reaction
- Undergoes Suzuki-Miyaura cross-coupling
- Appearance: Solid, white to off-white
- Target: Biochemical assay reagents
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.