Learn More
Medchemexpress LLC 2-Naphthylboronic acid | 32316-92-0 | 171.99 | 500 MG
SDP
Catalog No. 5000445837
Encompass_Preferred
2-Naphthylboronic acid serves as a biochemical reagent for life science research. It can function as a biological material or an organic compound, making it versatile for various experimental setups. This compound is notably capable of undergoing silylation reactions and readily participates in Suzuki-Miyaura cross-coupling reactions, crucial for synthesizing complex molecules.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Capable of undergoing silylation reactions
  • Participates in Suzuki-Miyaura cross-coupling reactions
Catalog No. 50-004-45837 Supplier Medchemexpress LLC Supplier No. HYW002570S500MG
2-Naphthylboronic acid serves as a biochemical reagent for life science research. It can function as a biological material or an organic compound, making it versatile for various experimental setups. This compound is notably capable of undergoing silylation reactions and readily participates in Suzuki-Miyaura cross-coupling reactions, crucial for synthesizing complex molecules.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Capable of undergoing silylation reactions
  • Participates in Suzuki-Miyaura cross-coupling reactions