Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Naphthylmethyl)Zinc Bromide Solution

Catalog No. 49902150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-902-150ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-902-150ML Supplier Sigma Aldrich Supplier No. 49902150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2-Naphthylmethyl)zinc bromide is an organozinc halide, which is typically used as a reagent in Negishi cross-coupling reactions. In this process, the organozinc halide reacts with organic halides or triflates to form a carbon-carbon bond in the presence of a Pd catalyst.(2-Naphthylmethyl)zinc bromide can be used as a reagent for the synthesis of: 4-(5,6-dimethoxy-2-(naphthalen-2-ylmethyl)pyridin-3-yl)benzonitrile from 4-(2-bromo-5,6-dimethoxypyridin-3-yl)benzonitrile using Pd catalyst. (E)-stilbene, (2,2'sa-[1,4-phenylenedi-(1E)-2,1-ethenediyl]bis[naphthalene]) from 1,4-benzenedicarboxaldehyde in the presence of Pd catalyst and a silylating agent.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 130792-45-9
Melting Point 90°C to 94°C
Molecular Formula C6H7BrO2
Linear Formula C6H7BrO2
MDL Number MFCD00216512
Quantity 5 g
Molecular Weight (g/mol) 191.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.