Learn More
Abcam 2-NBD-Glucose, Fluorescent glucose uptake probe, 1MG
Catalog No. 502583240
Encompass_Preferred
MW 342.26 g/mol. Fluorescently-labeled deoxyglucose analog, used to directly monitor glucose uptake by living cells and tissues. Also used as a topical contrast reagent for the detection of neoplasia. Uptake can be monitored by real-time confocal, high-resolution or wide-field fluorescence microscopy and flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3240 Supplier Abcam Supplier No. AB1462001MG
MW 342.26 g/mol. Fluorescently-labeled deoxyglucose analog, used to directly monitor glucose uptake by living cells and tissues. Also used as a topical contrast reagent for the detection of neoplasia. Uptake can be monitored by real-time confocal, high-resolution or wide-field fluorescence microscopy and flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement