Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-NBDG 100mg | 186689-07-6 | 100 MG
SDP

Catalog No. 5000229177
Encompass_Preferred

2-NBDG is a fluorescent D-glucose analog used to monitor glucose uptake in living cells and tissues by fluorescence-based assays. Supplied as a powder, it is typically prepared as aqueous stock solutions for cell-based experiments.

  • Fluorescent glucose uptake probe suitable for live-cell assays.
  • Excitation ~467 nm; emission ~542 nm.
  • Supplied as a powder soluble in water and PBS.
  • Typical supplier-reported purity ≥97%-98% (varies by vendor).
  • Molecular weight 342.26 g/mol; CAS 186689-07-6.

Catalog No. 50-002-29177 Supplier Medchemexpress LLC Supplier No. HY116215100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-NBDG is a fluorescent D-glucose analog used to monitor glucose uptake in living cells and tissues by fluorescence-based assays. Supplied as a powder, it is typically prepared as aqueous stock solutions for cell-based experiments.

  • Fluorescent glucose uptake probe suitable for live-cell assays.
  • Excitation ~467 nm; emission ~542 nm.
  • Supplied as a powder soluble in water and PBS.
  • Typical supplier-reported purity ≥97%-98% (varies by vendor).
  • Molecular weight 342.26 g/mol; CAS 186689-07-6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.