Learn More
Medchemexpress LLC HY-116215 10mg Medchemexpress, 2-NBDG CAS:186689-07-6 Purity:>98%
SDP
Catalog No. 501969409
Encompass_Preferred
Medchemexpress, HY-116215 10mg 2-NBDG CAS:186689-07-6 2-NBDG is a fluorescent indicator for direct glucose uptake measurement and also is an indicator of cell viability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9409 Supplier Medchemexpress LLC Supplier No. HY11621510MG
Medchemexpress, HY-116215 10mg 2-NBDG CAS:186689-07-6 2-NBDG is a fluorescent indicator for direct glucose uptake measurement and also is an indicator of cell viability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.