Learn More
Medchemexpress LLC 2-NBDG 25mg | 186689-07-6 | 342.26 | C12H14N4O8 | 25 MG
SDP
Catalog No. 5000229084
Encompass_Preferred
2-NBDG is a fluorescently labeled deoxyglucose analog used to directly monitor glucose uptake in living cells and tissues. It is excitable at 488 nm and produces environment-sensitive fluorescence suitable for real-time confocal, high-resolution, or wide-field fluorescence microscopy and for flow cytometry. Supplied as a 25 mg solid with high reported purity.
  • Directly reports cellular glucose uptake in live-cell assays.
  • Compatible with flow cytometry and fluorescence microscopy.
  • Excitable at 488 nm (argon laser).
  • Environment-sensitive fluorescence for intracellular localization studies.
  • High reported purity for reliable experimental results.
Catalog No. 50-002-29084 Supplier Medchemexpress LLC Supplier No. HY11621525MG
2-NBDG is a fluorescently labeled deoxyglucose analog used to directly monitor glucose uptake in living cells and tissues. It is excitable at 488 nm and produces environment-sensitive fluorescence suitable for real-time confocal, high-resolution, or wide-field fluorescence microscopy and for flow cytometry. Supplied as a 25 mg solid with high reported purity.
  • Directly reports cellular glucose uptake in live-cell assays.
  • Compatible with flow cytometry and fluorescence microscopy.
  • Excitable at 488 nm (argon laser).
  • Environment-sensitive fluorescence for intracellular localization studies.
  • High reported purity for reliable experimental results.