Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-NBDG 50mg | 186689-07-6 | 342.26 g/mol | C12H14N4O8 | 50 MG
SDP

Catalog No. 5000226443
Encompass_Preferred

2-NBDG is a fluorescent 2-deoxyglucose analog used to monitor glucose uptake in living cells and tissues. It is used as a fluorescent probe in real-time confocal, high-resolution, and wide-field fluorescence microscopy, in flow cytometry, and as a topical contrast reagent for neoplasia detection.

  • Fluorescent probe that reports glucose uptake in live cells and tissues.
  • Compatible with confocal, wide-field fluorescence microscopy, and flow cytometry.
  • Aqueous solubility: H2O 5 MG/ML (14.61 mM) with ultrasonic/warming and heating to 60°C; PBS 3.33 MG/ML with the same treatment.
  • Storage: protect from light; store at -20°C. In solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Molecular formula C12H14N4O8; molecular weight 342.26 g/mol.

Catalog No. 50-002-26443 Supplier Medchemexpress LLC Supplier No. HY11621550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-NBDG is a fluorescent 2-deoxyglucose analog used to monitor glucose uptake in living cells and tissues. It is used as a fluorescent probe in real-time confocal, high-resolution, and wide-field fluorescence microscopy, in flow cytometry, and as a topical contrast reagent for neoplasia detection.

  • Fluorescent probe that reports glucose uptake in live cells and tissues.
  • Compatible with confocal, wide-field fluorescence microscopy, and flow cytometry.
  • Aqueous solubility: H2O 5 MG/ML (14.61 mM) with ultrasonic/warming and heating to 60°C; PBS 3.33 MG/ML with the same treatment.
  • Storage: protect from light; store at -20°C. In solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Molecular formula C12H14N4O8; molecular weight 342.26 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.