Learn More
Apexbio Technology LLC 2-NBDG Glucose Uptake Assay Kit(Synonyms: 2-NBDG Glucose Assay Kit, Fluorescent Glucose Uptake Kit), 500T.
SDP
Catalog No. 5000061192
Encompass_Preferred
2-NBDG Glucose Uptake Assay Kit utilizes the fluorescently labeled glucose analog 2-NBDG (2-N-(7-nitrobenz-2-oxa-1 3-diazol-4-yl)amino-2-deoxyglucose) to quantify glucose uptake in cells 2-NBDG enters cells via glucose transporters (GLUTs) and undergoes phosphorylation at the C-6 position producing 2-NBDG-6-phosphate thus becoming intracellularly trapped This property enables precise measurement of cellular glucose uptake at the single-cell level through fluorescence analysis Relevant in metabolic pathway research and conditions including cancer diabetes and obesity this assay provides a rapid sensitive and non-radioactive approach to monitoring glucose metabolism
Catalog No. 50-000-61192 Supplier Apexbio Technology LLC Supplier No. K2212500
2-NBDG Glucose Uptake Assay Kit utilizes the fluorescently labeled glucose analog 2-NBDG (2-N-(7-nitrobenz-2-oxa-1 3-diazol-4-yl)amino-2-deoxyglucose) to quantify glucose uptake in cells 2-NBDG enters cells via glucose transporters (GLUTs) and undergoes phosphorylation at the C-6 position producing 2-NBDG-6-phosphate thus becoming intracellularly trapped This property enables precise measurement of cellular glucose uptake at the single-cell level through fluorescence analysis Relevant in metabolic pathway research and conditions including cancer diabetes and obesity this assay provides a rapid sensitive and non-radioactive approach to monitoring glucose metabolism