Learn More
Abcam 2-NBDG Glucose Uptake Assay Kit, 50 Tests
Supplier:  Abcam AB28784550TEST
Encompass_Preferred
2-NBDG Glucose Uptake Assay Kit ab287845 uses the fluorescent glucose analog 2-NBDG to quantify the rate of cellular glucose uptake. Individual kit components also available for purchase with a minimum order of 20 units. Contact us to discuss your needs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5914