Learn More
Chemglass Life Sciences 100mL 2-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 1-SN 10/30 Outer Angled 20°
SDP
Catalog No. CG152001
Encompass_Preferred
100mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 10/30 Outer Angled 20¿. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and angled side neck. Flasks with 10/30 side neck are for use with CG-3506 thermometers. The 29/26 center neck flasks have a groove tooled just below the grind to aid in clamping of the flask.
Catalog No. CG152001 Supplier Chemglass Life Sciences Supplier No. CG152001
100mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 10/30 Outer Angled 20¿. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and angled side neck. Flasks with 10/30 side neck are for use with CG-3506 thermometers. The 29/26 center neck flasks have a groove tooled just below the grind to aid in clamping of the flask.