Learn More
Cayman Chemical 2 NEthylperfluoroctnsulf 10mg
SDP
Catalog No. 502914979
Encompass_Preferred
A perfluoroalkyl substance; has been found in wastewater samples; associated with decreased plasma levels of maternal and fetal free T4
Catalog No. 50-291-4979 Supplier Cayman Chemical Supplier No. 3736310 MG
A perfluoroalkyl substance; has been found in wastewater samples; associated with decreased plasma levels of maternal and fetal free T4