Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. N084125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N084125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N084125G Supplier TCI America Supplier No. N084125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2267-23-4
Molecular Formula C7H5F3N2O3
Molecular Weight (g/mol) 222.12
MDL Number MFCD00042326
InChI Key YCGFVAPIBALHRT-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethoxy aniline, 2-amino-5-trifluoromethoxy nitrobenzene, benzenamine, 2-nitro-4-trifluoromethoxy, 1-amino-2-nitro-4-trifluoromethoxy benzene, 2-nitro-4-trifluoromethoxy phenylamine, 4-amino-3-nitro-alpha,alpha,alpha-trifluoroanisole, pubchem8514, acmc-1cnwp, ksc496g9b, 2-nitro4-trifluoromethoxyaniline
PubChem CID 2775772
IUPAC Name 2-nitro-4-(trifluoromethoxy)aniline
SMILES NC1=CC=C(OC(F)(F)F)C=C1[N+]([O-])=O

Specifications

Melting Point 63°C
Quantity 25 g
Formula Weight 222.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.