Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-nitro-5-methylbenzoic acid | 3113-72-2 | MFCD00007368 | 99.9% | 181.15 g/mol | C8H7NO4 | 1 KG
SDP

Catalog No. 5000303198
Encompass_Preferred

5-Methyl-2-nitrobenzoic acid is an organic laboratory reagent (2-nitro-5-methylbenzoic acid) used as a building block and intermediate in organic synthesis and life-science research. It is provided as a high-purity solid with documented physical and analytical data for laboratory use.

  • High purity: 99.9%
  • Appearance: solid, white to light yellow
  • Molecular formula: C8H7NO4; molecular weight: 181.15 g/mol
  • Solubility: soluble in DMSO (100 mg/mL) with ultrasonic assistance
  • Storage: powder stable at -20°C (long term) and 4°C (short term)
  • Includes analytical documentation: COA, SDS, HNMR, RP-HPLC available

Catalog No. 50-003-03198 Supplier Medchemexpress LLC Supplier No. HYW0164501KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-nitrobenzoic acid is an organic laboratory reagent (2-nitro-5-methylbenzoic acid) used as a building block and intermediate in organic synthesis and life-science research. It is provided as a high-purity solid with documented physical and analytical data for laboratory use.

  • High purity: 99.9%
  • Appearance: solid, white to light yellow
  • Molecular formula: C8H7NO4; molecular weight: 181.15 g/mol
  • Solubility: soluble in DMSO (100 mg/mL) with ultrasonic assistance
  • Storage: powder stable at -20°C (long term) and 4°C (short term)
  • Includes analytical documentation: COA, SDS, HNMR, RP-HPLC available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.