Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-nitro-5-methylbenzoic acid | 3113-72-2 | MFCD00007368 | 99.9% | 181.15 g/mol | C8H7NO4 | 100 G
SDP

Catalog No. 5000303766
Encompass_Preferred

5-Methyl-2-nitrobenzoic acid (2-nitro-5-methylbenzoic acid) is an aromatic carboxylic acid used as a biochemical reagent and as a building block in organic synthesis. Supplied as a solid, it is suitable for research, analytical standards, and intermediate synthesis in medicinal chemistry and analytical workflows.

  • High purity: 99.90%
  • Molecular weight: 181.15 g/mol
  • Chemical formula: C8H7NO4
  • Appearance: white to light yellow solid
  • Soluble in DMSO (100 mg/mL; may require sonication)
  • Storage: powder at -20°C (3 years) or 4°C (2 years)

Catalog No. 50-003-03766 Supplier Medchemexpress LLC Supplier No. HYW016450100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methyl-2-nitrobenzoic acid (2-nitro-5-methylbenzoic acid) is an aromatic carboxylic acid used as a biochemical reagent and as a building block in organic synthesis. Supplied as a solid, it is suitable for research, analytical standards, and intermediate synthesis in medicinal chemistry and analytical workflows.

  • High purity: 99.90%
  • Molecular weight: 181.15 g/mol
  • Chemical formula: C8H7NO4
  • Appearance: white to light yellow solid
  • Soluble in DMSO (100 mg/mL; may require sonication)
  • Storage: powder at -20°C (3 years) or 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.