Learn More
Medchemexpress LLC 2-nitro-6-(6-(4-trifluoromethoxy)phenylpyridin-3-yl)methoxy-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine | 1257426-19-9 | MFCD28009369 | 99.7% | 436.34 | C19H15F3N4O5 | 50MG
SDP
Catalog No. 5000236400
Encompass_Preferred
TBA-354 is a nitroimidazole-derived antitubercular research compound supplied as a solid for laboratory use. It is described as a potent agent active against Mycobacterium tuberculosis H37Rv and certain monoresistant strains, and is intended for in vitro and preclinical research only.
  • High purity (99.74%) confirmed by certificate of analysis.
  • Molecular formula C19H15F3N4O5 and molecular weight 436.34 g·mol⁻¹.
  • Supplied as a 50 mg solid for research use.
  • Nitroimidazole-class compound with activity against M. tuberculosis.
  • Certificate of analysis and analytical data provided for quality control.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-36400 Supplier Medchemexpress LLC Supplier No. HY1248550MG
TBA-354 is a nitroimidazole-derived antitubercular research compound supplied as a solid for laboratory use. It is described as a potent agent active against Mycobacterium tuberculosis H37Rv and certain monoresistant strains, and is intended for in vitro and preclinical research only.
  • High purity (99.74%) confirmed by certificate of analysis.
  • Molecular formula C19H15F3N4O5 and molecular weight 436.34 g·mol⁻¹.
  • Supplied as a 50 mg solid for research use.
  • Nitroimidazole-class compound with activity against M. tuberculosis.
  • Certificate of analysis and analytical data provided for quality control.
  • For research use only; not for human or veterinary use.