Learn More
Sigma Aldrich 2-Nitro-alpha-toluenesulfonyl fluoride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- (+/-)-1-(2-furyl)ethanol [(+/-)-2-furyl methyl carbinol] is a furan derivative
- The synthesis of 4-hydroxy-2-methylcyclopent-2-en-1-one from 2-furyl methyl carbinol has been reported.(+/-)-1-(2-furyl)ethanol (racemic 1-(2-furyl)ethanol) may be used in the synthesis of 1-acetoxy-1-[2-furyl]ethan
Specifications
Specifications
| Molecular Formula | C7H6FNO4S |
| Linear Formula | C7H6FNO4S |
| MDL Number | MFCD22571069 |
| Quantity | 10 mL |
| Synonym | 2-Nitro-alpha-toluenesulfonyl fluoride |
| Molecular Weight (g/mol) | 219.19 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.